missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypochlorite solution, Reagent Grade, available chlorine 10-15 %, Honeywell™

Product Code. 16663954 Shop All Honeywell Research Chemicals Products
Click to view available options
:
25 L
This item is not returnable. View return policy

Product Code. 16663954

Brand: Honeywell Research Chemicals 42504425L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 111°C
Chemical Name or Material Sodium hypochlorite solution
Melting Point -30°C to -20°C
CAS 7681-52-9
Color Clear
CAS Min % >85%
CAS Max % ≤ 95%
Quantity 25 L
Linear Formula NaClO
Molecular Weight (g/mol) 74.44 g/mol
Grade Reagent
Density 1.206 g/cm3 (25°C)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.