missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite, Sterile, Made with USP Purified Water, 5% Solution, BAKER™, J.T. Baker™

Product Code. 15548794 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 gL
Unit Size:
3.8 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15548794 1 gL 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15548794 Supplier Avantor J.T.Baker Supplier No. P00503
  • ONLINE EXCLUSIVE PRICE
    347.63 €
    291.54 € / 3.8 L
    Save 56.09 €
    16% Off
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

A grade of chemicals for laboratory use

Specifica

CAS 7681-52-9
Physical Form Liquid
Quantity 1 gL
Molecular Formula ClNaO
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
SMILES [O-]Cl.[Na+]
IUPAC Name sodium;hypochlorite
Molecular Weight (g/mol) 74.439
PubChem CID 23665760
ChEBI CHEBI:32146
Formula Weight 74.44
Percent Purity 5%
Grade Pure
Specific Gravity 1L = 1.11kg
Chemical Name or Material Sodium Hypochlorite, Sterile, Made with USP Purified Water
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.