missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite, Technical, Solution, Self Indicating, Fisher Chemical

Product Code. 10090480 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 L
Packaging:
HDPE plastic bottle
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10090480 5 L HDPE plastic bottle 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10090480 Supplier Fisher Chemical Supplier No. S/5041/21
  • ONLINE EXCLUSIVE PRICE
    574.00 €
    568.00 € / 5 L
    Save 6.00 €
    1% Off
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

Melting Point -16°C
CAS 7681-52-9
Color Yellow
pH 11
Physical Form Liquid
Quantity 5 L
Molecular Formula ClNaO
MDL Number 11120
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
SMILES [O-]Cl.[Na+]
IUPAC Name sodium;hypochlorite
Molecular Weight (g/mol) 74.439
PubChem CID 23665760
ChEBI CHEBI:32146
Packaging HDPE plastic bottle
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium hypochlorite
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.