missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypophosphite monohydrate, 97+%, water ca 12-17%

Product Code. 11425383
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425383 2500 g 2500g
11415383 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425383 Supplier Thermo Scientific Alfa Aesar Supplier No. A13385.0E
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10039-56-2 | H4NaO3P | 105.99 g/mol

Sodium hypophosphite monohydrate is used for electroless nickel plating agent and water treatment agent. It is also used in petroleum fields. Further, it is used as a food additive. In addition, it reduces nickel ions in solution to metallic nickel on metal substrates as well as on plastic substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10039-56-2
Molecular Formula H4NaO3P
Molecular Weight (g/mol) 105.99
MDL Number MFCD06252379
InChI Key PLZNPHDJGFDNRM-UHFFFAOYSA-M
Synonym Sodium phosphinite monohydrate;
IUPAC Name sodium hydrate oxophosphinite
SMILES O.[Na+].[O-][PH2]=O

Specifications

Physical Form Solid
Quantity 2500 g
Percent Purity ≥97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.