missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypophosphite monohydrate, for analysis

Product Code. 11974281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11974281 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11974281 Supplier Thermo Scientific Acros Supplier No. 222790051
  • ONLINE EXCLUSIVE PRICE
    441.00 €
    428.00 € / 5kg
    Save 13.00 €
    2% Off
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10039-56-2 | H4NaO3P | 105.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 90.0°C
CAS 10039-56-2
Quantity 5 kg
Molecular Formula H4NaO3P
Linear Formula NaH2PO2·H2O
MDL Number MFCD06252379
Synonym sodium hypophosphite hydrate, sodium phosphinate hydrate, sodium phosphenite hydrate, sodium hypophosphite en grade, sodium hydrate hypophosphite, sodium hypophosphite monohydrate 250g, phosphinic acid sodium salt monohydrate, sodium hypophosphite monohydrate, phosphonous acid monosodium salt monohydrate, sodium hypophosphite monohydrate, puriss
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: 33.0 g/100 g ethylene glycol (25°C), 9.7 g/100 g propylene glycol (25°C), freely soluble in glycerol and boiling alcohol, slightly soluble in absolute alcohol, insoluble in ether - soluble in cold
InChI Key PLZNPHDJGFDNRM-UHFFFAOYSA-M
SMILES O.[Na+].[O-][PH2]=O
IUPAC Name sodium hydrate phosphinate
Molecular Weight (g/mol) 105.99
PubChem CID 23708894
Formula Weight 105.99
Grade Analytical
Chemical Name or Material Sodium hypophosphite monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.