missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypophosphite Monohydrate, puriss., Honeywell

Product Code. 15630770 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15630770 1 kg 1kg
15640770 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15630770 Supplier Honeywell Fluka Supplier No. 044151KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10039-56-2
Molecular Formula H4NaO3P
Molecular Weight (g/mol) 105.99
MDL Number MFCD06252379
InChI Key PLZNPHDJGFDNRM-UHFFFAOYSA-M
Synonym sodium hypophosphite hydrate, sodium phosphinate hydrate, sodium phosphenite hydrate, sodium hypophosphite en grade, sodium hydrate hypophosphite, sodium hypophosphite monohydrate 250g, phosphinic acid sodium salt monohydrate, sodium hypophosphite monohydrate, phosphonous acid monosodium salt monohydrate, sodium hypophosphite monohydrate, puriss
PubChem CID 23708894
IUPAC Name sodium hydrate phosphinate
SMILES O.[Na+].[O-][PH2]=O

Specifications

Quantity 1 kg
Linear Formula NaH2PO2 · H2O
UN Number NONH for all modes of transport
Formula Weight 105.99g/mol
Percent Purity 99 to 103%
Grade puriss.
Chemical Name or Material Sodium Hypophosphite Monohydrate, Meets analytical specification of DAC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.