missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypophosphite monohydrate, puriss., meets analytical specification of DAC, 99-103%, Honeywell

Product Code. 15650770 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Fiberboard Box
Unit Size:
50kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15650770 50 kg Fiberboard Box 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15650770 Supplier Honeywell Fluka Supplier No. 0441550KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 10039-56-2
CAS Max % 100%
Quantity 50 kg
Assay Percent Range 99 to 103%
Molecular Formula H4NaO3P
Linear Formula NaH2PO2 · H2O
MDL Number MFCD06252379
UN Number NONH for all modes of transport
Synonym sodium hypophosphite hydrate, sodium phosphinate hydrate, sodium phosphenite hydrate, sodium hypophosphite en grade, sodium hydrate hypophosphite, sodium hypophosphite monohydrate 250g, phosphinic acid sodium salt monohydrate, sodium hypophosphite monohydrate, phosphonous acid monosodium salt monohydrate, sodium hypophosphite monohydrate, puriss
InChI Key PLZNPHDJGFDNRM-UHFFFAOYSA-M
SMILES O.[Na+].[O-][PH2]=O
IUPAC Name sodium hydrate phosphinate
Molecular Weight (g/mol) 105.99
PubChem CID 23708894
Formula Weight 105.99g/mol
Percent Purity 99 to 103%
Grade puriss.
Packaging Fiberboard Box
Chemical Name or Material Sodium hypophosphite monohydrate, puriss., meets analytical specification of DAC, 99-103%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.