missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodide, 99+%, pure, anhydrous

Product Code. 10154640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
10 kg
Unit Size:
10kg
1kg
2.5kg
250g
5g
Product Code. Quantity unitSize
10154640 5 g 5g
10206980 250 g 250g
10616252 1 kg 1kg
10689253 2.5 kg 2.5kg
15264674 10 kg 10kg
5 options
This item is not returnable. View return policy

Product Code. 10154640

Brand: Thermo Scientific Acros 203180050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7681-82-5 | INa | 149.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.89
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Boiling Point 1300.0°C
Melting Point 661.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99% min. (on dried substance) (Argentometry)
Linear Formula NaI
Fieser 01,1087; 02,384; 03,267; 04,456; 06,543; 07,338; 10,365; 11,488; 12,29; 13,43; 14,89; 16,86
Merck Index 15,8765
Solubility Information Solubility in water: 184 g/100mL (25°C). Other solubilities: soluble in alcohol, glycerol and acetone
Formula Weight 149.89
Percent Purity ≥99%
Grade Pure
Loss on Drying 3% max. (1 g, 105°C, 3 hrs)
Packaging Glass bottle
Specific Gravity 3.66
Density 3.6600g/mL
Chemical Name or Material Sodium iodide, Anhydrous
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium iodide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.