missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Iodide, Certified AR for Analysis, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Specifications
Specifications
| Boiling Point | 1300°C |
| Melting Point | 661°C |
| Color | White |
| pH | 6 to 9 |
| Physical Form | Powder/Solid |
| Quantity | 500 g |
| Formula Weight | 149.89g/mol |
| Packaging | Plastic powder jar |
| Vapor Pressure | 1.3mbar 767 |
Safety and Handling
Product Identifier
- Sodium iodide
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P273-Avoid release to the environment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction