missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide, puriss., Solstice
BIOPHARMA

Product Code. 15690670 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 500 g
6 x 1 kg
Unit Size:
100g
1kg
500g
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15690670 500 g 500g
15670670 100 g 100g
15680670 1 kg 1kg
15610680 6 x 500 g Pack of 6
15600680 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15690670 Supplier Honeywell-Fluka Supplier No. 03129500G
  • ONLINE EXCLUSIVE PRICE
    195.00 €
    175.88 € / 500g
    Save 19.12 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Quantity 500 g
Linear Formula NaI
UN Number 3077
Formula Weight 149.89g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Packaging Plastic Bottle
Density Vapor Density: >1 (vs air)
Chemical Name or Material Sodium Iodide, Meets analytical specification of Ph.Eur. BP, USP
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium iodide
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.