missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodide, ultra dry, 99.98% (metals basis)

Product Code. 11345757
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11345757 25 g 25g
11335757 100 g 100g
11355757 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 11345757

Brand: Thermo Scientific Alfa Aesar 043447.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as iodine supplement, expectorant or in its radioactive (I-131) form as an diagnostic aid, particularly for thyroid function tests. sodium Iodide are used in photography and as analytical reagents. It is also used in the measurement of the energy of gamma rays, also used in organic synthesis. | CAS: 7681-82-5 | INa | 149.894 g/mol

It is used as iodine supplement, expectorant or in its radioactive (I-131) form as an diagnostic aid, particularly for thyroid function tests. sodium Iodide are used in photography and as analytical reagents. It is also used in the measurement of the energy of gamma rays, also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Boiling Point 1300°C to 1304°C
Melting Point 651°C
Physical Form Beads
Quantity 25 g
Merck Index 14,8631
Solubility Information It is soluble in water,alcohol,and glycerin.
Formula Weight 149.89
Percent Purity 99.98%
Odor Odorless
Packaging Ampule
Refractive Index 1.7745
Sensitivity Hygroscopic
Density 3.67 g/mL
Assay (metals basis)
Chemical Name or Material Sodium iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.