missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iron EDTA, TRC

Product Code. 30415226
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415226 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415226 Supplier Toronto Research Chemicals Supplier No. TRCS63499050G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Iron EDTA
CAS 15708-41-5
Molecular Formula C10 H12 Fe N2 O8 . Na
Quantity 50 g
Synonym Ferrate(1-)','[[N,N'-1,2-ethanediylbis[N-[(carboxy-κO)methyl]glycinato-κN,κO]](4-)]-','sodium (1:1)','(OC-6-21)-, Ferrate(1-)','[(ethylenedinitrilo)tetraacetato]-','sodium (8CI), Ferrate(1-)','[[N,N'-1,2-ethanediylbis[N-(carboxymethyl)glycinato]](4-)-N,N',O,O',ON,ON']-','sodium','(OC-6-21)-, Ferrate(1-)','[[N,N'-1,2-ethanediylbis[N-[(carboxy-κO)methyl]glycinato-κN,κO]](4-)]-','sodium','(OC-6-21)- (9CI), Sodium [(ethylenedinitrilo)tetraacetato]ferrate(III) (6CI,7CI), AL 3037A, EDTA iron (III) sodium salt, Edathamil monosodium ferric salt, Ethylenediaminetetraacetic acid ferric monosodium salt, Fe-EDTA, Ferisan, Ferrazone, Ferric sodium EDTA, Ferric sodium edetate, Ferric sodium ethylenediaminetetraacetate, Ferrostrane, Ferrostrene, Ferroxx, Iron monosodium EDTA, Iron sodium EDTA, Iron sodium ethylenediaminetetraacetate, Iron sodium ethylenediaminetetraacetate (1:1:1), Monosodium ferric EDTA, NSC 5237, Sequestrene NaFe, Sodium (N,N,N',N'-ethylenediaminetetraacetato)ferrate(1-), Sodium (ethylenediaminetetraacetato)ferrate(1-), Sodium feredetate, Sodium ferric EDTA, Sodium ferric ethylenediaminetetraacetate, Sodium iron EDTA, Sodium iron(III) ethylenediaminetetraacetate, Sybron, Sytron
InChI Formula InChI=1S/C10H16N2O8.Fe.Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;/q;+3;+1/p-4
SMILES [Na+].O=C1CN23CCN45CC(=O)[O-][Fe+3]24([O-]C(=O)C5)([O-]C(=O)C3)[O-]1
Molecular Weight (g/mol) 367.05
Formula Weight 366.9841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.