missing translation for 'onlineSavingsMsg'
Learn More

Sodium L-lactate, 98+%

Product Code. 11353556
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353556 5 g 5g
11363556 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353556 Supplier Thermo Scientific Alfa Aesar Supplier No. L14500.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 867-56-1 | C3H5NaO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium L-lactate is used as a food additive, preservative, acidity regulator and bulking agent. It finds application in shampoo and liquid soaps and other related products. It acts as an effective humectant and moisturizer. It is used in the treatment of arrhythmias.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 867-56-1
Molecular Formula C3H5NaO3
MDL Number MFCD00066576
Synonym sodium l-lactate, sodium-l-lactate, sodium lactate, l, sodium s-2-hydroxypropanoate, unii-p2y1c6m9ps, sodium s-lactate, sodium l-+-lactate, p2y1c6m9ps, propanoic acid, 2-hydroxy-, monosodium salt, 2s, l-+-lactic acid sodium salt

Specifications

Melting Point ∼160°C
Quantity 5 g
Beilstein 4567087
Solubility Information Soluble in water.
Formula Weight 112.06
Percent Purity ≥98%
Sensitivity Hygroscopic
Chemical Name or Material Sodium L-lactate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.