missing translation for 'onlineSavingsMsg'
Learn More

Sodium Lactate Solution, Extra Pure, SLR, about 60% (w/w) Solution, Fisher Chemical

Product Code. 10213970 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10213970 500 mL Amber glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10213970 Supplier Fisher Chemical Supplier No. S/5161/08
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Boiling Point 140°C
Melting Point 17.2°C
CAS 72-17-3
Color Yellow
pH 7
Physical Form Viscous Liquid
Quantity 500 mL
Molecular Formula C3H5NaO3
MDL Number 66576
Synonym sodium lactate, lactic acid sodium salt, monosodium lactate, sodium dl-lactate, lacolin, per-glycerin, lactic acid, monosodium salt, sodium alpha-hydroxypropionate, monosodium 2-hydroxypropanoate, propanoic acid, 2-hydroxy-, monosodium salt
InChI Key NGSFWBMYFKHRBD-UHFFFAOYSA-M
SMILES CC(C(=O)[O-])O.[Na+]
IUPAC Name sodium;2-hydroxypropanoate
Molecular Weight (g/mol) 112.06
PubChem CID 23666456
ChEBI CHEBI:75228
Packaging Amber glass bottle
Vapor Pressure 14mmHg at 20°C
Density Vapor Density: 0.7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.