missing translation for 'onlineSavingsMsg'
Learn More

Sodium mercury amalgam, ca. 5% sodium

Product Code. 10628984 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10628984 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10628984 Supplier Thermo Scientific Acros Supplier No. 388590100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 11110-52-4 | HgNa | 223.582 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Color Gray
CAS 11110-52-4
Quantity 10 g
Packaging Glass bottle
Form Chunk
Molecular Formula HgNa
MDL Number MFCD00210362
Merck Index 15, 8713
Synonym sodium amalgam, sodium mercury amalgam, sodium mercury, sodium-amalgam, mercury sodium, sodium mercury amalgam, beads, na 10 %, sodium mercury amalgam, beads trace metals basis, na 5 %, sodium mercury amalgam, beads trace metals basis, na 20 %
InChI Key MJGFBOZCAJSGQW-UHFFFAOYSA-N
SMILES [Na].[Hg]
IUPAC Name mercury;sodium
Molecular Weight (g/mol) 223.582
PubChem CID 9837419
Chemical Name or Material Sodium mercury amalgam
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium mercury amalgam, ca. 5% sodium
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H360D-May damage the unborn child.
  • H330-Fatal if inhaled.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.