missing translation for 'onlineSavingsMsg'
Learn More

Sodium meta-Bisulfite, Multi-Compendial, J.T. Baker™

Product Code. 15528144 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15528144 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15528144 Supplier Avantor J.T.Baker Supplier No. 355105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7681-57-4
Quantity 2.5 kg
Molecular Formula Na2O5S2
MDL Number MFCD00167602
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O
IUPAC Name disodium (sulfinatooxy)sulfinate
Molecular Weight (g/mol) 190.09
PubChem CID 656671
Formula Weight 190.11
Grade Multi-Compendial
Packaging Poly Bottle
Specific Gravity 1L = 1.48kg
Chemical Name or Material Sodium meta-Bisulfite
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium metabisulphite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium metabisulphite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.