missing translation for 'onlineSavingsMsg'
Learn More

Sodium meta-Periodate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15568164 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Bottle
Unit Size:
125g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15568164 500 g Poly Bottle 500g
15172017 125 g Poly Bottle 125g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15568164 Supplier Avantor J.T.Baker Supplier No. 375601
  • ONLINE EXCLUSIVE PRICE
    1345.00 €
    1294.94 € / 500g
    Save 50.06 €
    3% Off
 Restricted Item
Estimated Shipment: 09-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
MDL Number MFCD00003534
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate
PubChem CID 23667635
ChEBI CHEBI:75226
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Quantity 500 g
IUPAC Name sodium;periodate
Formula Weight 213.89
Percent Purity 99.9%
Grade Meets ACS reagent chemical requirements
Packaging Poly Bottle
Specific Gravity 1L = 3.87kg
Chemical Name or Material Sodium meta-Periodate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.