missing translation for 'onlineSavingsMsg'
Learn More

Sodium (Meta)Periodate, Solstice

Product Code. 15653010
Change view
Click to view available options
Quantity:
100 g
500 g
6 x 500 g
Unit Size:
100g
500g
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15653010 6 x 500 g Pack of 6
15633010 100 g 100g
15643010 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15653010 Supplier Honeywell-Fluka Supplier No. 303236X500G
  • ONLINE EXCLUSIVE PRICE
    2185.00 €
    1972.25 € / Pack of 6
    Save 212.75 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.8%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
MDL Number MFCD00003534
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate
PubChem CID 23667635
ChEBI CHEBI:75226
IUPAC Name sodium;periodate
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Quantity 6 x 500 g
Linear Formula NaIO4
UN Number UN3085
Formula Weight 213.89
Percent Purity ≥99.8%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium (Meta)Periodate, Reag. Ph. Eur. reag. ISO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.