missing translation for 'onlineSavingsMsg'
Learn More

Sodium metabisulfite, 97+%, ACS reagent

Product Code. 10361351 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
2.5kg
250g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10361351 5 g Glass Bottle 5g
10444235 250 g Plastic bottle 250g
10301311 1 kg Plastic bottle 1kg
10267903 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10361351 Supplier Thermo Scientific Acros Supplier No. 419580050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7681-57-4 | Na2O5S2 | 190.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
PubChem CID 656671
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

Melting Point 150.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 5 g
Assay Percent Range 97% min. (Iodimetry)
Linear Formula Na2S2O5
Merck Index 15, 8770
Solubility Information Solubility in water: 540g/L (20°C). Other solubilities: freely soluble in glycerol, slightly soluble in alcohol
Formula Weight 190.1
Percent Purity ≥97%
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Sodium metabisulfite
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium metabisulfite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.