missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Metabisulfite NF/FCC Granular, Macron Fine Chemicals™
Specifications
Specifications
| CAS | 7681-57-4 |
| Physical Form | Granules |
| Quantity | 2.5 kg |
| Molecular Formula | Na2O5S2 |
| MDL Number | MFCD00167602 |
| Synonym | sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis |
| InChI Key | LDTLADDKFLAYJA-UHFFFAOYSA-L |
| SMILES | [Na+].[Na+].[O-]S(=O)OS([O-])=O |
| IUPAC Name | disodium (sulfinatooxy)sulfinate |
| Molecular Weight (g/mol) | 190.09 |
| Show More |
Safety and Handling
Product Identifier
- Sodium metabisulphite
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: Sodium metabisulphite
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?