missing translation for 'onlineSavingsMsg'
Learn More

Sodium metabisulfite, SO{2} 58.5% min

Product Code. 11347698
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347698 500 g 500g
11337698 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347698 Supplier Thermo Scientific Alfa Aesar Supplier No. 033375.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7681-57-4 ∣ Na2O5S2 ∣ 190.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
PubChem CID 656671
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

Melting Point >300°C
Color White to Yellow
pH 3.5 to 5.0
Physical Form Granules
Quantity 500 g
Merck Index 14,8638
Solubility Information Freely soluble in water, glycerol; Slightly soluble in alcohol
Formula Weight 190.1
Grade Ungraded
Concentration or Composition (by Analyte or Components) SO2 58.5% min.
Odor Like rotten eggs
Sensitivity Air and moisture sensitive
Density 1.480 g/mL
Chemical Name or Material Sodium metabisulfite
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.