missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metabisulfite, TRC

Product Code. 17952825
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17952825 50 g 50g
17930725 10 g 10g
17934425 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17952825 Supplier Toronto Research Chemicals Supplier No. TRCS66706050G
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-57-4
Molecular Formula 2 Na . O5 S2
Molecular Weight (g/mol) 190.11
Synonym Sodium metabisulfite, sodium disulfite
SMILES [Na+].[Na+].[O-]S(=O)S(=O)(=O)[O-]

Specifications

Quantity 50 g
InChI Formula InChI=1S/2Na.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2
Formula Weight 189.8983
Chemical Name or Material Sodium Metabisulfite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.