missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metasilicate Pentahydrate, Technical, Fisher Chemical™

Product Code. 10746111 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 kg
3 kg
Unit Size:
1kg
3kg
This item is not returnable. View return policy

Product Code. 10746111

Brand: Fisher Chemical S/5360/60

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sodium Metasilicate Pentahydrate, Technical, Fisher Chemical, Quantity: 3kg, CAS: 10213-79-3, White, Melting Point: 72 deg.C, Molecular Formula: H10Na2O8Si, Molecular Weight: 212.137, Packaging: Plastic powder jar, pH: 12.6

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10213-79-3
Molecular Formula H10Na2O8Si
Molecular Weight (g/mol) 212.137
MDL Number 149176
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
Synonym sodium metasilicate pentahydrate, silicic acid, disodium salt, pentahydrate, disodium oxosilanediolate pentahydrate, silicic acid h2sio3 , disodium salt, pentahydrate, 2na.so3i.5h2o, sodium meta-silicate pentahydrate, sodium silicate solution water glass, disodium dioxido oxo silane pentahydrate, sodium metasilicate pentahydrate, tech., disodium oxosilanebis olate pentahydrate
PubChem CID 61495
IUPAC Name disodium;dioxido(oxo)silane;pentahydrate
SMILES O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Melting Point 72°C
Color White
pH 12.6
Physical Form Solid
Quantity 1 kg
Formula Weight 212.74g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Sodium metasilicate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium metasilicatepentahydrate, Sodiummetasilicate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.