missing translation for 'onlineSavingsMsg'
Learn More

Sodium metatungstate hydrate, crystalline

Product Code. 10423075 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 10423075

Brand: Thermo Scientific Acros 304090010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 652154-12-6 | H2Na6O40W12·xH2O | 2986.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652154-12-6
Molecular Formula H2Na6O40W12·xH2O
Molecular Weight (g/mol) 2986.13
MDL Number MFCD00150663
Synonym Sodium polytungstate Hydrate

Specifications

Color White to Yellow
Physical Form Crystalline Powder or Beads
Quantity 1 kg
Linear Formula Na6[H2W12O40]·xH2O
Solubility Information Solubility in water: >1000g/L (20°C)
Formula Weight 2986.13
Percent Purity ≥85% (WO3)
Packaging Glass bottle
Chemical Name or Material Sodium metatungstate hydrate
compliance-icons compliance-icons
Product Identifier
  • Sodium metatungstate hydrate, crystalline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.