missing translation for 'onlineSavingsMsg'
Learn More

Sodium metavanadate, 96%

Product Code. 10185213 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10185213 1 kg Glass bottle 1kg
10105020 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10185213 Supplier Thermo Scientific Acros Supplier No. 269210010
  • ONLINE EXCLUSIVE PRICE
    550.00 €
    513.00 € / 1kg
    Save 37.00 €
    6% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13718-26-8 ∣ NaO3V ∣ 121.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13718-26-8
Molecular Formula NaO3V
Molecular Weight (g/mol) 121.93
MDL Number MFCD00011125
InChI Key CMZUMMUJMWNLFH-UHFFFAOYSA-N
Synonym sodium metavanadate, sodium vanadate v, vanadic acid, monosodium salt, sodium vanadate meta, sodium oxido dioxo vanadium, sodium trioxovanadate 1-, metawanadan sodowy polish, vanadate vo31-, sodium, monosodium trioxovanadate 1-, ccris 6881
PubChem CID 4148882
ChEBI CHEBI:75221
IUPAC Name sodium vanadiumoylolate
SMILES [Na+].[O-][V](=O)=O

Specifications

Melting Point 600°C
Color Beige-Yellow to White or Green
Physical Form Powder
Quantity 1 kg
Packaging Glass bottle
Assay Percent Range 96%
Linear Formula NaVO3
Merck Index 15,8774
Solubility Information Solubility in water: soluble
Formula Weight 121.93
Percent Purity 96%
Grade Ungraded
Chemical Name or Material Sodium metavanadate, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.