missing translation for 'onlineSavingsMsg'
Learn More

Sodium Methanethiolate (~20% in Water), TRC

Product Code. 30180390
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180390 1 g 1g
30180207 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180390 Supplier Toronto Research Chemicals Supplier No. TRCS6448001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5188-07-8
Molecular Formula C H3 S . Na
Molecular Weight (g/mol) 70.09
Synonym Methanethiol, sodium salt (1:1), Methanethiol, sodium salt (8CI,9CI), Sodium, (methylthio)- (7CI), (Methylsulfanyl)sodium, Methyl mercaptan sodium salt, Methylthiosodium, Sodium methanesulfenate, Sodium methanethiolate, Sodium methyl mercaptan, Sodium methyl sulfide, Sodium methylmercaptide, Sodium methylthiolate, Sodium thiomethoxide, Sodium thiomethylate
IUPAC Name sodium;methanethiolate
SMILES [Na+].C[S-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/CH4S.Na/c1-2;/h2H,1H3;/q;+1/p-1
Formula Weight 69.9853
Concentration or Composition (by Analyte or Components) Sodium Methanethiolate ~20%
Chemical Name or Material Sodium Methanethiolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.