missing translation for 'onlineSavingsMsg'
Learn More

Sodium methoxide, ACS reagent, 0.5M solution in methanol, AcroSeal™

Product Code. 11936161 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11936161 800 mL AcroSeal™ Glass bottle 800mL
11906161 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11936161 Supplier Thermo Scientific Acros Supplier No. 445888000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124-41-4 ∣ CH3NaO ∣ 54.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-41-4 , 67-56-1
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.02
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Boiling Point 65.0°C
Melting Point -98.0°C
Physical Form Solution
Quantity 800 mL
Linear Formula CH3ONa
Flash Point 11°C
Merck Index 15,8775
Solubility Information Solubility in water: reacts
Formula Weight 54.02
Grade ACS/Reagent
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.8
Density 0.8000g/mL
Chemical Name or Material Sodium methoxide, 0.5M solution in methanol, AcroSeal™
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.