missing translation for 'onlineSavingsMsg'
Learn More

Sodium methoxide, ca 30% w/w in methanol

Product Code. 11434937
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434937 2500 mL 2500mL
11424937 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434937 Supplier Thermo Scientific Alfa Aesar Supplier No. B21872.0F
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 124-41-4 | CH3NaO | 54.024 g/mol

Sodium methoxide, ca 30% w/w in methanol is used in the preparation of high-spin hexanuclear Mn(III) cluster, [NaMn6 (OMe)12 (dbm)6]+. It is useful in the synthesis of pharmaceuticals and agrochemicals. It serves as an initiator of anionic addition polymerization with ethylene oxide, forming a polyether with high molecular weight.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium methoxide, ca 30% w/w in methanol is used in the preparation of high-spin hexanuclear Mn(III) cluster, [NaMn6 (OMe)12 (dbm)6]+. It is useful in the synthesis of pharmaceuticals and agrochemicals. It serves as an initiator of anionic addition polymerization with ethylene oxide, forming a polyether with high molecular weight.

Solubility
Miscible with ethanol, fats and esters. Immiscible with hydrocarbons and organic solvents.

Notes
Air, moisture and heat sensitive. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents, alkali metals, chlorinated solvents, water and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-41-4
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.024
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Melting Point <6°C
Density 0.969
Boiling Point 93°C
Flash Point 16°C (61°F)
Odor Alcohol-like
Linear Formula NaOCH3
Quantity 2500 mL
UN Number UN1289
Beilstein 3592982
Sensitivity Air and moisture sensitive
Merck Index 14,8643
Solubility Information Miscible with ethanol,fats and esters. Immiscible with hydrocarbons and organic solvents.
Formula Weight 54.02
Concentration or Composition (by Analyte or Components) ≈30% w/w in methanol
Chemical Name or Material Sodium methoxide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium methoxide, ca 30% w/w in methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: methanol, sodium methylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.