missing translation for 'onlineSavingsMsg'
Learn More

Sodium Methoxide, TRC

Product Code. 30414990
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Product Code. Quantity unitSize
30414990 1 kg 1kg
1 options
This item is not returnable. View return policy

Product Code. 30414990

Brand: Toronto Research Chemicals TRCS6454001KG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium methoxide
CAS 124-41-4
Molecular Formula C H3 O . Na
Quantity 1 kg
Synonym Methanol','sodium salt (1:1), Methanol','sodium salt (8CI,9CI), Sodium methoxide (7CI), Feldalat NM, Methoxysodium, SM 24, SM 28, Sodium methanolate, Sodium methoxylate, Sodium methylate, Natriummethylat
InChI Formula InChI=1S/CH3O.Na/c1-2;/h1H3;/q-1;+1
SMILES [Na+].C[O-]
IUPAC Name sodium;methanolate
Molecular Weight (g/mol) 54.02
Formula Weight 54.0082
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.