missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate, Dihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15578164 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15578164 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15578164 Supplier Avantor J.T.Baker Supplier No. 376401
  • ONLINE EXCLUSIVE PRICE
    518.66 €
    469.78 € / 500g
    Save 48.88 €
    9% Off
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 10102-40-6
Quantity 500 g
Molecular Formula H4MoNa2O6
MDL Number MFCD00149170
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
Molecular Weight (g/mol) 241.96
PubChem CID 16211258
ChEBI CHEBI:75213
Formula Weight 241.95
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 3.28kg
Chemical Name or Material Sodium Molybdate, Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.