missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-dodecyl sulfate (SDS), 20% aq. soln.

Product Code. 15465639
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465639 250 mL 250mL
15475639 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465639 Supplier Thermo Scientific Alfa Aesar Supplier No. J63394.AK
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

Sodium n-dodecyl sulfate 20% aq. solution is used as an anionic surfactant. It is also used as fat emulsifier, a wetting agent, a detergent in cosmetics, pharmaceuticals and toothpastes. It also serves as a research tool in protein biochemistry. It is mainly used in detergents for laundry with many cleaning applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium n-dodecyl sulfate 20% aq. solution is used as an anionic surfactant. It is also used as fat emulsifier, a wetting agent, a detergent in cosmetics, pharmaceuticals and toothpastes. It also serves as a research tool in protein biochemistry. It is mainly used in detergents for laundry with many cleaning applications.

Solubility
Miscible with ethanol.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color Colorless
Physical Form Liquid
Concentration 20% aq. soln.
Quantity 250 mL
Linear Formula CH3(CH2)11OSO3Na
Solubility Information Miscible with ethanol.
IUPAC Name sodium;dodecyl sulfate
Formula Weight 288.38
Chemical Name or Material Sodium n-dodecyl sulfate (SDS)
compliance-icons
Product Identifier
  • Sodium n-dodecyl sulfate (SDS), 20% aq. soln.
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium lauryl sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.