missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-octadecyl sulfate, 98% (dry wt.), water <7%

Product Code. 11477297
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477297 25 g 25g
11457297 1 g 1g
11467297 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11477297 Supplier Thermo Scientific Alfa Aesar Supplier No. A15042.14
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1120-04-3 | C18H37NaO4S | 372.54 g/mol

Sodium n-octadecyl sulfate is used in the formulation of hair care products including shampoo, skin cleansers, and other skin care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium n-octadecyl sulfate is used in the formulation of hair care products including shampoo, skin cleansers, and other skin care products.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-04-3
Molecular Formula C18H37NaO4S
Molecular Weight (g/mol) 372.54
MDL Number MFCD00007469
InChI Key NWZBFJYXRGSRGD-UHFFFAOYSA-M
Synonym sodium stearyl sulfate, octadecyl sulfate sodium salt, sodium octadecyl sulfate, octadecyl sodium sulfate, sulfuric acid, monooctadecyl ester, sodium salt, sodium octadecyl sulphate, sodium monostearyl sulfate, unii-t4eww0y4ej, sodium n-octadecyl sulfate, sodium monooctadecyl sulfate
PubChem CID 23673604
IUPAC Name sodium;octadecyl sulfate
SMILES CCCCCCCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 176°C to 180°C
Linear Formula CH3(CH2)17OSO3Na
Quantity 25 g
Beilstein 3640871
Solubility Information Soluble in water.
Formula Weight 372.55
Percent Purity 98%
Assay (dry wt.), water <7%
Chemical Name or Material Sodium n-octadecyl sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.