missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nafcillin Monohydrate, TRC

Product Code. 17963075
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963075 1 g 1g
17903645 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17963075 Supplier Toronto Research Chemicals Supplier No. TRCS6458001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7177-50-6
Molecular Formula C21 H21 N2 O5 S . Na . H2 O
Molecular Weight (g/mol) 454.47
Synonym Nafcillin Sodium Monohydrate, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(2-ethoxy-1-naphthalenyl)carbonyl]amino]-3,3-dimethyl-7-oxo-, sodium salt, hydrate (1:1:1), (2S,5R,6R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-(2-ethoxy-1-naphthamido)-3,3-dimethyl-7-oxo-, monosodium salt, monohydrate (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(2-ethoxy-1-naphthalenyl)carbonyl]amino]-3,3-dimethyl-7-oxo-, monosodium salt, monohydrate, (2S,5R,6R)- (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(2-ethoxy-1-naphthalenyl)carbonyl]amino]-3,3-dimethyl-7-oxo-, monosodium salt, monohydrate, [2S-(2α,5α,6β)]-, (2S,5R,6R)-6-[[(2-Ethoxy-1-naphthalenyl)carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid sodium salt monohydrate, Nafcillin sodium hydrate, Sodium 6-(2-ethoxy-1-naphthamido)penicillanate monohydrate, Sodium nafcillin monohydrate
IUPAC Name sodium;(2S,5R,6R)-6-[(2-ethoxynaphthalene-1-carbonyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate
SMILES O.[Na+].CCOc1ccc2ccccc2c1C(=O)N[C@H]3[C@H]4SC(C)(C)[C@@H](N4C3=O)C(=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H22N2O5S.Na.H2O/c1-4-28-13-10-9-11-7-5-6-8-12(11)14(13)17(24)22-15-18(25)23-16(20(26)27)21(2,3)29-19(15)23;;/h5-10,15-16,19H,4H2,1-3H3,(H,22,24)(H,26,27);;1H2/q;+1;/p-1/t15-,16+,19-;;/m1../s1
Formula Weight 454.1175
Percent Purity >95%
Chemical Name or Material Nafcillin sodium salt monohydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.