missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium nitrite, 97+%, ACS reagent
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
2 kg
5 kg
Unit Size:
2kg
5g
5kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Physical Form | Crystalline Powder and/or Crystals |
| Quantity | 5 g |
| Assay Percent Range | 97+% |
| Linear Formula | NaNO2 |
| Fieser | 01,1097; 02,386; 04,459; 06,547; 09,432; 11,491; 13,170; 16,86 |
| Merck Index | 15, 8779 |
| Formula Weight | 69 |
| Percent Purity | ≥97.0% |
| Grade | ACS Reagent |
| Show More |
Safety and Handling
Product Identifier
- Sodium nitrite
- Danger
- Oxidising solid Category 3
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P273-Avoid release to the environment.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction