missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrite, 99%, extra pure, contains an anticaking reagent

Product Code. 10742295 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
10kg
1kg
2.5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10742295 1 kg Plastic bottle 1kg
10012580 2.5 kg Plastic bottle 2.5kg
10370494 10 kg Plastic Drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10742295 Supplier Thermo Scientific Acros Supplier No. 196620010
  • ONLINE EXCLUSIVE PRICE
    71.30 €
    66.90 € / 1kg
    Save 4.40 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7632-00-0 | NNaO2 | 69.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium;nitrite
SMILES [Na+].[O-]N=O

Specifications

Color Yellow
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 99%
Linear Formula NaNO2
Fieser 01,1097; 02,386; 04,459; 06,547; 09,432; 11,491; 13,170; 16,86
Merck Index 15, 8779
Formula Weight 69
Percent Purity ≥98.5%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Sodium nitrite, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.