missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrite, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10437660 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10437660 100 g 100g
10784581 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10437660 Supplier JT Baker Supplier No. 0297.0100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Exceeds ACS Specifications. Meets Reagent Specifications for testing USP/NF monographs

Chemical Identifiers

Molecular Formula NaNO2
Molecular Weight (g/mol) 69
MDL Number MFCD00011118

Specifications

Boiling Point 320°C (1013 hPa)
Melting Point 284°C
Quantity 100 g
Assay Percent Range min. 97.0%
Density 2.168 g/cm³ (20°C)
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H301-Toxic if swallowed.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.