missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, puriss. p.a. ACS Reagent, Solstice

Product Code. 15633430
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Unit Size:
100g
1kg
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15633430 100 g 100g
15653430 250 g 250g
15663430 500 g 500g
15643430 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15633430 Supplier Honeywell-Fluka Supplier No. 31443100G
  • ONLINE EXCLUSIVE PRICE
    38.00 €
    35.00 € / 100g
    Save 3.00 €
    7% Off
 Restricted Item
Estimated Shipment: 28-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium;nitrite
SMILES [Na+].[O-]N=O

Tekniske data

Quantity 100 g
Linear Formula NaNO2
UN Number 1500
Formula Weight 69g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Ignition Point 914°F
Chemical Name or Material Sodium Nitrite, Reag. Ph. Eur.
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • sodium nitrate
Signalord
  • Danger
Farekategori
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Oxidising solid Category 3
Faresætning
  • H272-May intensify fire; oxidiser.
  • H301-Toxic if swallowed.
  • H400-Very toxic to aquatic life.
Sikkerhedssætning
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplerende oplysninger
  • MIXTURE LIST-Contain: sodium nitrate
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.