missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium nitrite solution, 0.1M, Honeywell™
Click to view available options
Quantity:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
CAS | 7632-00-0 |
Quantity | 1 L |
Molecular Formula | NNaO2 |
Linear Formula | NaNO2 |
MDL Number | MFCD00011118 |
UN Number | NONH for all modes of transport |
Synonym | sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts |
InChI Key | LPXPTNMVRIOKMN-UHFFFAOYSA-M |
SMILES | [Na+].[O-]N=O |
IUPAC Name | sodium;nitrite |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'