missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitroferricyanide dihydrate, reagent ACS,meets the requirements of Reag.Ph.Eur.

Product Code. 10321651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10321651 100 g Glass bottle 100g
10239470 25 g Glass bottle 25g
10381691 500 g Glass bottle 500g
10107103 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10321651 Supplier Thermo Scientific Acros Supplier No. 424361000
  • ONLINE EXCLUSIVE PRICE
    123.00 €
    115.00 € / 100g
    Save 8.00 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13755-38-9 | C5H4FeN6Na2O3 | 297.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Color Bordeaux
Physical Form Crystals
Quantity 100 g
Packaging Glass bottle
Specific Gravity 1.72
Density 1.7200g/mL
Linear Formula Na2Fe(CN)5NO·2H2O
Merck Index 15,878
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
Formula Weight 297.94
Percent Purity 99.0 to 102.0%
Grade ACS Reagent
Chemical Name or Material Sodium nitroferricyanide dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium nitroferricyanide dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contain: Sodium nitroferricyanide dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.