missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroferricyanide(Iii) Dihydrate, Solstice

Product Code. 15692270 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15692270 500 g 500g
15602280 5 g 5g
15682270 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15692270 Supplier Honeywell-Fluka Supplier No. 228710500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Quantity 500 g
Linear Formula Na2[Fe(CN)5NO] · 2H2O
UN Number 3288
IUPAC Name disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
Formula Weight 297.95g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Sodium Nitroferricyanide(Iii) Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.