missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroferricyanide Dihydrate, ACS Reagent Grade, Ricca Chemical

Product Code. 31750302
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31750302 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31750302 Supplier Ricca Chemical Company Supplier No. RDCS0700100B1
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 13755-38-9
CAS Min % 98.0
CAS Max % 100.0
Physical Form Fine Crystals or Crystalline Powder
Quantity 100 g
Packaging White Poly Wide Mouth Bottle
Molecular Formula C5H4FeN6Na2O3
MDL Number MFCD00149192
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
IUPAC Name disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
Molecular Weight (g/mol) 297.95
PubChem CID 11953895
Percent Purity 99 to 102%
Grade ACS
Chemical Name or Material Sodium Nitroferricyanide, Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.