missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroprusside Dihydrate, Certified AR for Analysis, Fisher Chemical

Product Code. 11488842 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11488842 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11488842 Supplier Fisher Chemical Supplier No. S/5720/48
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

100g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

CAS 13755-38-9
Color Red
pH 5
Physical Form Crystalline Solid
Quantity 100 g
Molecular Formula C5H4FeN6Na2O3
MDL Number MFCD00149192
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
IUPAC Name disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
Molecular Weight (g/mol) 297.95
PubChem CID 11953895
Formula Weight 297.94g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Sodium nitroferricyanide dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contain: Sodium nitroferricyanide dihydrate, Sodium nitroferricyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.