missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroprusside Dihydrate, purum p.a., ≥98% (AT), ACS reagent, Honeywell™

Product Code. 15616850 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15616850 25 g 25g
15606850 100 g 100g
15626850 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15616850 Supplier Honeywell Research Chemicals Supplier No. 7178025G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥98.0% (AT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Quantity 25 g
Linear Formula Na2[Fe(CN)5NO] · 2H2O
UN Number UN3288
IUPAC Name disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
Formula Weight 297.95g/mol
Percent Purity ≥98% (AT)
Chemical Name or Material Sodium nitroprusside dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.