missing translation for 'onlineSavingsMsg'
Learn More

Sodium oleate, Honeywell Fluka™

Product Code. 16635783 Shop All Honeywell Fluka Products
Click to view available options
:
15KG POLY DRUM
This item is not returnable. View return policy

Product Code. 16635783

missing translation for 'mfr': Honeywell Fluka 26125H15KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

≥82% fatty acids (as oleic acid) basis, powder

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 143-19-1
Physical Form Solid
Quantity 15 kg
Flash Point 149 °C
PubChem CID 44135713
Percent Purity ≥82%
Packaging Poly Drum
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.