missing translation for 'onlineSavingsMsg'
Learn More

Sodium orthovanadate, 100mM aq. soln.

Product Code. 15403009
Change view
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15403009 50 mL 50mL
15413009 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15403009 Supplier Thermo Scientific Alfa Aesar Supplier No. J60191.AD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13721-39-6 | Na3O4V | 183.907 g/mol

Sodium orthovanadate, is used as an inhibitor of several phosphatases and kinases such as protein tyrosine phosphatases (PTPs), alkaline phosphatase (ALP), and ATPase. It also acts as an inhibitor of adenosine kinase (AK) and phosphofructokinase (PFK). It also suppresses p53-mediated apoptosis by transcription-dependent and -independent pathways.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium orthovanadate, is used as an inhibitor of several phosphatases and kinases such as protein tyrosine phosphatases (PTPs), alkaline phosphatase (ALP), and ATPase. It also acts as an inhibitor of adenosine kinase (AK) and phosphofructokinase (PFK). It also suppresses p53-mediated apoptosis by transcription-dependent and -independent pathways.

Solubility
It is soluble in water (50 mg/ml, )

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13721-39-6
Molecular Formula Na3O4V
Molecular Weight (g/mol) 183.907
MDL Number MFCD00003511
InChI Key IHIXIJGXTJIKRB-UHFFFAOYSA-N
Synonym sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
PubChem CID 61671
ChEBI CHEBI:35607
IUPAC Name trisodium;trioxido(oxo)vanadium
SMILES [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 50 mL
Linear Formula Na3VO4
Solubility Information It is soluble in water (50mg/ml,)
Formula Weight 183.94
Concentration or Composition (by Analyte or Components) 100mM aq. soln.
Odor Odorless
Chemical Name or Material Sodium orthovanadate
Is Sodium orthovanadate, 100 mM aq. soln. (Cat. No. J60191.xx) activated?

Sodium orthovanadate, 100mM aq. soln. (Cat. No. J60191.xx) is activated and suitable for use as a phosphatase inhibitor. Activation was achieved by boiling and adjusting the pH to 10.0.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.