missing translation for 'onlineSavingsMsg'
Learn More

Sodium orthovanadate, 99.9% (metals basis)

Product Code. 11304919
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304919 500 g 500g
11384909 25 g 25g
11394909 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11304919 Supplier Thermo Scientific Alfa Aesar Supplier No. 081104.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

inhibitor of Protein Tyrosine Phosphatase and Alkaline Phosphate | CAS: 13721-39-6 | Na3O4V | 183.907 g/mol

Inhibitor of protein tyrosine phosphatase and alkaline phosphate Sodium orthovanadate is used in protein analysis in molecular biology and acts as an inhibitor of protein tyrosine phosphatases, alkaline phosphatases and kinases. It is also used to prepare the stop solution in dephosphorylation assay of insulin receptor kinase. It serves as a reagent in the development of matrix Chip that utilizes surface-immobilized antibodies. Further, it is utilized for the preservation of protein tyrosyl phosphorylation state in cells and cell lysates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13721-39-6
Molecular Formula Na3O4V
Molecular Weight (g/mol) 183.907
MDL Number MFCD00003511
InChI Key IHIXIJGXTJIKRB-UHFFFAOYSA-N
Synonym sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
PubChem CID 61671
ChEBI CHEBI:35607
IUPAC Name trisodium;trioxido(oxo)vanadium
SMILES [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Melting Point 850°C to 866°C
Color White
Physical Form Granules
Quantity 500 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in water.
Formula Weight 183.94
Odor Odorless
Chemical Name or Material Sodium orthovanadate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.