missing translation for 'onlineSavingsMsg'
Learn More

Sodium oxalate, 98.5%, extra pure

Product Code. 10442211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10442211 100 g Plastic bottle 100g
10052710 500 g Plastic bottle 500g
10391614 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10442211 Supplier Thermo Scientific Acros Supplier No. 207721000
  • ONLINE EXCLUSIVE PRICE
    30.30 €
    29.40 € / 100g
    Save 0.90 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 62-76-0 | C2Na2O4 | 134.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium;oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Melting Point 250.0°C to 270.0°C
Color White
Loss on Drying 0.1% max. (105°C, 2 hrs)
Assay Percent Range 98.5%
Packaging Plastic bottle
Linear Formula NaO2CCO2Na
Quantity 100 g
Merck Index 15, 8781
Solubility Information Solubility in water: 37g/L (20°C). Other solubilities: 6.3g/100mL water (100°C),insoluble in ethanol and ether
Formula Weight 134
Percent Purity 98.5%
Grade Extra Pure
Insoluble Matter 0.2% max. (in water)
Physical Form Fine Powder
Chemical Name or Material Sodium oxalate, pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.