missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, 99.8+%, Certified AR for Analysis, Fisher Chemical™

Product Code. 12616777 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12616777 500 g 500g
12606777 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12616777 Supplier Fisher Chemical Supplier No. S/5840/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium;oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Melting Point 250°C
Color White
pH 8
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 134g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Sodium oxalate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.