missing translation for 'onlineSavingsMsg'
Learn More

Sodium oxalate, ACS reagent

Product Code. 12377183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12377183 500 g Plastic bottle 500g
12367183 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12377183 Supplier Thermo Scientific Acros Supplier No. 446965000
  • ONLINE EXCLUSIVE PRICE
    115.00 €
    107.00 € / 500g
    Save 8.00 €
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 62-76-0 ∣ C2Na2O4 ∣ 134.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium;oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Especificaciones

Melting Point 250.0°C to 270.0°C
Physical Form Crystalline Powder
Quantity 500 g
Packaging Plastic bottle
Linear Formula NaO2CCO2Na
Merck Index 15,8781
Solubility Information Solubility in water: 37g/L (20°C). Other solubilities: 6.3 g/100mL water (100°C), insoluble in ethanol and ether
Formula Weight 134
Grade ACS/Reagent
Chemical Name or Material Sodium oxalate
compliance-icons
Nombre del producto
  • Sodium oxalate
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Otros peligros
  • MIXTURE LIST-Contain: Ethanedioic acid, disodium salt

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.