missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, TRC

Product Code. 30007431
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007431 50 g 50g
30010229 5 g 5g
30009782 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007431 Supplier Toronto Research Chemicals Supplier No. TRCS69934850G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2O4.2Na
Molecular Weight (g/mol) 134
Synonym Disodium oxalate;
IUPAC Name disodium;oxalate
SMILES [Na+].[Na+].[O-]C(=O)C(=O)[O-]

Specifications

Quantity 50 g
InChI Formula InChI=1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2
Formula Weight 133.9592
Chemical Name or Material Sodium Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.